4-Aminohippuric acid, 99%, Thermo Scientific Chemicals
4-Aminohippuric acid, 99%, Thermo Scientific Chemicals
4-Aminohippuric acid, 99%, Thermo Scientific Chemicals
4-Aminohippuric acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Aminohippuric acid, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
10 g
25 g
100 g
Packaging:
View all
Plastic bottle
Catalog number ACR153805000
Price (MYR)
12,418.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
12,418.00
Add to cart
Chemical Identifiers
CAS1176-74-5
IUPAC Namepotassium 2,6-dibromo-4-[(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)[2-(ethoxycarbonyl)phenyl]methyl]benzen-1-olate
Molecular FormulaC22H13Br4KO4
InChI KeyWCIQBKUTYDIBJC-UHFFFAOYSA-M
SMILES[K+].CCOC(=O)C1=CC=CC=C1C(C1=CC(Br)=C([O-])C(Br)=C1)=C1C=C(Br)C(=O)C(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Powder
Appearance (Color)Yellow to red
UV spectrophotometry>=80.0 %