4-Aminohippuric acid, 99%, Thermo Scientific Chemicals
4-Aminohippuric acid, 99%, Thermo Scientific Chemicals
4-Aminohippuric acid, 99%, Thermo Scientific Chemicals
4-Aminohippuric acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Aminohippuric acid, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
10 g
25 g
500 g
Packaging:
View all
Plastic bottle
Catalog number ACR153801000
Price (MYR)
2,711.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
2,711.00
Add to cart
Chemical Identifiers
CAS127-79-7
IUPAC Name4-amino-N-(4-methylpyrimidin-2-yl)benzene-1-sulfonamide
Molecular FormulaC11H12N4O2S
InChI KeyQPPBRPIAZZHUNT-UHFFFAOYSA-N
SMILESCC1=CC=NC(NS(=O)(=O)C2=CC=C(N)C=C2)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥98.0%
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder