4-Aminohippuric acid, 99%, Thermo Scientific Chemicals
4-Aminohippuric acid, 99%, Thermo Scientific Chemicals
4-Aminohippuric acid, 99%, Thermo Scientific Chemicals
4-Aminohippuric acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Aminohippuric acid, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
10 g
100 g
500 g
Packaging:
View all
Plastic bottle
Catalog number ACR153800250
Price (MYR)
944.00
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
944.00
Add to cart
Chemical Identifiers
CAS100-63-0
IUPAC Namehydrogen phenylhydrazine chloride
Molecular FormulaC6H9ClN2
InChI KeyJOVOSQBPPZZESK-UHFFFAOYSA-N
SMILES[H+].[Cl-].NNC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange to brown
Assay (Non-aqueous acid-base Titration)≥96.0% (UK-sourced material)
CommentMaterial sourced in UK and US
FormLiquid or fused solid
Assay from Supplier's CofA≥96.0% (US-sourced material)