4-Aminohippuric acid, 99%, Thermo Scientific Chemicals
4-Aminohippuric acid, 99%, Thermo Scientific Chemicals
4-Aminohippuric acid, 99%, Thermo Scientific Chemicals
4-Aminohippuric acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Aminohippuric acid, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
100 g
500 g
Packaging:
View all
Glass Bottle
Catalog number ACR153800100
Price (MYR)
619.00
Add to cart
Quantity:
10 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
619.00
Add to cart
Specifications
Chemical Name or Material4-Aminohippuric acid
CAS61-78-9
CAS Min %98.5
Health Hazard 1Warning
CAS Max %100.0