D(+)-Malic acid, 98%, Thermo Scientific Chemicals
D(+)-Malic acid, 98%, Thermo Scientific Chemicals
D(+)-Malic acid, 98%, Thermo Scientific Chemicals
D(+)-Malic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
D(+)-Malic acid, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number ACR153500010
Price (MYR)
462.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
462.00
Add to cart
Specifications
Chemical Name or MaterialD(+)-Malic acid
CAS636-61-3
Melting Point99.0°C to 104.0°C
Health Hazard 1Danger