2-Deoxy-D-ribose, 99%
2-Deoxy-D-ribose, 99%
2-Deoxy-D-ribose, 99%
2-Deoxy-D-ribose, 99%
Thermo Scientific Chemicals
2-Deoxy-D-ribose, 99%
CAS: 533-67-5 | C5H10O4 | 134.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number ACR153280050
Price (MYR)
700.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
700.00
Add to cart
Specifications
Chemical Name or Material2-Deoxy-D-ribose
Name Note99%
CAS533-67-5
CAS Min %98.5
CAS Max %100.0