1-Bromopentane, 98%, Thermo Scientific Chemicals
1-Bromopentane, 98%, Thermo Scientific Chemicals
1-Bromopentane, 98%, Thermo Scientific Chemicals
1-Bromopentane, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Bromopentane, 98%, Thermo Scientific Chemicals
CAS: 110-53-2 | C5H11Br | 151.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
250 mL
Packaging:
Glass bottle
Catalog number ACR152520010
Price (MYR)
1,795.00
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,795.00
Add to cart
Specifications
Chemical Name or Material1-Bromopentane
Name Note98%
CAS110-53-2
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning