p-Anisic acid, 98%, Thermo Scientific Chemicals
p-Anisic acid, 98%, Thermo Scientific Chemicals
p-Anisic acid, 98%, Thermo Scientific Chemicals
p-Anisic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
p-Anisic acid, 98%, Thermo Scientific Chemicals
CAS: 100-09-4 | C8H8O3 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
100 g
Packaging:
Plastic bottle
Catalog number ACR152372500
Price (MYR)
641.00
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
641.00
Add to cart
Chemical Identifiers
CAS1310-53-8
IUPAC Namegermanedione
Molecular FormulaGeO2
InChI KeyYBMRDBCBODYGJE-UHFFFAOYSA-N
SMILESO=[Ge]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.0001% max.