p-Anisic acid, 98%, Thermo Scientific Chemicals
p-Anisic acid, 98%, Thermo Scientific Chemicals
p-Anisic acid, 98%, Thermo Scientific Chemicals
p-Anisic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
p-Anisic acid, 98%, Thermo Scientific Chemicals
CAS: 100-09-4 | C8H8O3 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
Packaging:
Plastic bottle
Catalog number ACR152371000
Price (MYR)
424.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
424.00
Add to cart
Specifications
Chemical Name or Materialp-Anisic acid
Name Note98%
CAS100-09-4
Melting Point181.0°C to 186.0°C
ColorBeige-Gray to White