2,2'-Biphenol, 99%, Thermo Scientific Chemicals
2,2'-Biphenol, 99%, Thermo Scientific Chemicals
2,2'-Biphenol, 99%, Thermo Scientific Chemicals
2,2'-Biphenol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,2'-Biphenol, 99%, Thermo Scientific Chemicals
CAS: 1806-29-7 | C12H10O2 | 186.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number ACR151991000
Price (MYR)
582.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
582.00
Add to cart
Chemical Identifiers
CAS51-55-8
IUPAC Name8-methyl-8-azabicyclo[3.2.1]octan-3-yl 3-hydroxy-2-phenylpropanoate
Molecular FormulaC17H23NO3
InChI KeyRKUNBYITZUJHSG-UHFFFAOYNA-N
SMILESCN1C2CCC1CC(C2)OC(=O)C(CO)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point114-118°C
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay from Supplier's CofA≥98.5
Water Content (Karl Fischer Titration)≤0.2%