2-Bromo-3'-methoxyacetophenone, 97%, Thermo Scientific Chemicals
2-Bromo-3'-methoxyacetophenone, 97%, Thermo Scientific Chemicals
2-Bromo-3'-methoxyacetophenone, 97%, Thermo Scientific Chemicals
2-Bromo-3'-methoxyacetophenone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Bromo-3'-methoxyacetophenone, 97%, Thermo Scientific Chemicals
CAS: 5000-65-7 | C9H9BrO2 | 229.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
10 g
Packaging:
Glass bottle
Catalog number ACR151910250
Price (MYR)
1,023.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,023.00
Add to cart
Specifications
Chemical Name or Material2-Bromo-3'-methoxyacetophenone
Name Note97%
CAS5000-65-7
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger