D(+)-alpha-Methylbenzylamine, 99+%, (99% ee), Thermo Scientific Chemicals
D(+)-alpha-Methylbenzylamine, 99+%, (99% ee), Thermo Scientific Chemicals
D(+)-alpha-Methylbenzylamine, 99+%, (99% ee), Thermo Scientific Chemicals
D(+)-alpha-Methylbenzylamine, 99+%, (99% ee), Thermo Scientific Chemicals
Thermo Scientific Chemicals
D(+)-alpha-Methylbenzylamine, 99+%, (99% ee), Thermo Scientific Chemicals
CAS: 3886-69-9 | C8H11N | 121.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number ACR151820250
Price (MYR)
554.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
554.00
Add to cart
Chemical Identifiers
CAS75-33-2
IUPAC Namepropane-2-thiol
Molecular FormulaC3H8S
InChI KeyKJRCEJOSASVSRA-UHFFFAOYSA-N
SMILESCC(C)S
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4240-1.4280 @ 20?C
Appearance (Color)Colorless
Assay (GC)≥97.5%
FormLiquid
Chiral derivatization reagent
RUO – Research Use Only