Bromophenol Blue, sodium salt, pure, water soluble, indicator
Bromophenol Blue, sodium salt, pure, water soluble, indicator
Bromophenol Blue, sodium salt, pure, water soluble, indicator
Bromophenol Blue, sodium salt, pure, water soluble, indicator
Thermo Scientific Chemicals
Bromophenol Blue, sodium salt, pure, water soluble, indicator
CAS: 62625-28-9 | C19H9Br4NaO5S | 691.942 g/mol
Have Questions?
Catalog NumberQuantity
ACR15135025025 g
Catalog number ACR151350250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS16630-52-7
SpecificationsSpecification SheetSpecification Sheet
Free acid (titration)≤2.5%
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.4755-1.4815 @ 20?C
FormLiquid
Assay (GC)≥94.0%
Indicator
RUO – Research Use Only