Bromophenol Blue, sodium salt, pure, water soluble, indicator
Bromophenol Blue, sodium salt, pure, water soluble, indicator
Bromophenol Blue, sodium salt, pure, water soluble, indicator
Bromophenol Blue, sodium salt, pure, water soluble, indicator
Thermo Scientific Chemicals
Bromophenol Blue, sodium salt, pure, water soluble, indicator
CAS: 62625-28-9 | C19H9Br4NaO5S | 691.942 g/mol
Have Questions?
Catalog NumberQuantity
ACR1513500505 g
Catalog number ACR151350050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS601-89-8
IUPAC Name2-nitrobenzene-1,3-diol
Molecular FormulaC6H5NO4
InChI KeyZLCPKMIJYMHZMJ-UHFFFAOYSA-N
SMILESOC1=CC=CC(O)=C1[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)Orange
Melting Point (clear melt)80-86?C
Assay (GC)≥97.5%
Indicator
RUO – Research Use Only