Crotonic acid, 98%, Thermo Scientific Chemicals
Crotonic acid, 98%, Thermo Scientific Chemicals
Crotonic acid, 98%, Thermo Scientific Chemicals
Crotonic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Crotonic acid, 98%, Thermo Scientific Chemicals
CAS: 107-93-7 | C4H6O2 | 86.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Plastic bottle
Catalog number ACR150875000
Price (MYR)
522.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
522.00
Add to cart
Specifications
Chemical Name or MaterialCrotonic acid
Name Note98%
CAS107-93-7
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement