tert-Butylhydroquinone, 97%, Thermo Scientific Chemicals
tert-Butylhydroquinone, 97%, Thermo Scientific Chemicals
tert-Butylhydroquinone, 97%, Thermo Scientific Chemicals
tert-Butylhydroquinone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
tert-Butylhydroquinone, 97%, Thermo Scientific Chemicals
CAS: 1948-33-0 | C10H14O2 | 166.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 g
500 g
Packaging:
View all
Glass Bottle
Catalog number ACR150820050
Price (MYR)
368.00
Add to cart
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
368.00
Add to cart
Chemical Identifiers
CAS90-44-8
IUPAC Name9,10-dihydroanthracen-9-one
Molecular FormulaC14H10O
InChI KeyRJGDLRCDCYRQOQ-UHFFFAOYSA-N
SMILESO=C1C2=CC=CC=C2CC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
CommentAbsorbance of reagent solution P.T.
CommentSensitivity to carbohydrates P.T.
Melting Pointno more than a 5? range including 156?
Solution Testin ethyl acetate P.T.