1-Butanethiol, 98%, Thermo Scientific Chemicals
1-Butanethiol, 98%, Thermo Scientific Chemicals
1-Butanethiol, 98%, Thermo Scientific Chemicals
1-Butanethiol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Butanethiol, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass bottle
Catalog number ACR150800010
Price (MYR)
1,121.00
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,121.00
Add to cart
Chemical Identifiers
CAS495-76-1
IUPAC Name(2H-1,3-benzodioxol-5-yl)methanol
Molecular FormulaC8H8O3
InChI KeyBHUIUXNAPJIDOG-UHFFFAOYSA-N
SMILESOCC1=CC=C2OCOC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
FormCrystals or powder or crystalline powder
Melting Point (clear melt)49.0-55.0?C
Assay (GC)≥97.5%