2-Methoxy-4-nitroaniline, 98%, Thermo Scientific Chemicals
2-Methoxy-4-nitroaniline, 98%, Thermo Scientific Chemicals
2-Methoxy-4-nitroaniline, 98%, Thermo Scientific Chemicals
2-Methoxy-4-nitroaniline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Methoxy-4-nitroaniline, 98%, Thermo Scientific Chemicals
CAS: 97-52-9 | C7H8N2O3 | 168.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number ACR150641000
Price (MYR)
452.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
452.00
Add to cart
Specifications
Chemical Name or Material2-Methoxy-4-nitroaniline
CAS97-52-9
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0