L(-)-Malic acid, 99%, Thermo Scientific Chemicals
L(-)-Malic acid, 99%, Thermo Scientific Chemicals
L(-)-Malic acid, 99%, Thermo Scientific Chemicals
L(-)-Malic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
L(-)-Malic acid, 99%, Thermo Scientific Chemicals
CAS: 97-67-6 | C4H6O5 | 134.087 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Plastic bottle
Catalog number ACR150595000
Price (MYR)
1,929.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,929.00
Add to cart
Chemical Identifiers
CAS7585-39-9
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)97.0% min.
Water Content (Karl Fischer Titration)14.0% max.
Trace Metals5ppm max.