L(-)-Malic acid, 99%, Thermo Scientific Chemicals
L(-)-Malic acid, 99%, Thermo Scientific Chemicals
L(-)-Malic acid, 99%, Thermo Scientific Chemicals
L(-)-Malic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
L(-)-Malic acid, 99%, Thermo Scientific Chemicals
CAS: 97-67-6 | C4H6O5 | 134.087 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Packaging:
Plastic bottle
Catalog number ACR150591000
Price (MYR)
578.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
578.00
Add to cart
Chemical Identifiers
CAS51-05-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white to almost white
Infrared spectrumConforms
Loss on drying=<0.5 % (1 g, 105°C)
Sulfated ash=<0.1 %
Appearance (Form)Crystalline powder or crystals