L(-)-Malic acid, 99%, Thermo Scientific Chemicals
L(-)-Malic acid, 99%, Thermo Scientific Chemicals
L(-)-Malic acid, 99%, Thermo Scientific Chemicals
L(-)-Malic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
L(-)-Malic acid, 99%, Thermo Scientific Chemicals
CAS: 97-67-6 | C4H6O5 | 134.087 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Catalog number ACR150590250
Price (MYR)
403.00
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
403.00
Add to cart
Chemical Identifiers
CAS71889-02-6
IUPAC Namesilanedione; trichloro(octadecyl)silane
Molecular FormulaC18H37Cl3O2Si2
InChI KeyVYTRPUAMVCWPLO-UHFFFAOYSA-N
SMILESO=[Si]=O.CCCCCCCCCCCCCCCCCC[Si](Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Carbon (C)>=20.7 %
Volatile matter=<4 % (110°C)
Specific surface area480 to 550 m²/g
Pore volume0.70 to 0.85 ml/g
Loading>=0.96 mmol/g (Molecular)