4-Bromobenzyl bromide, 98%, Thermo Scientific Chemicals
4-Bromobenzyl bromide, 98%, Thermo Scientific Chemicals
4-Bromobenzyl bromide, 98%, Thermo Scientific Chemicals
4-Bromobenzyl bromide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Bromobenzyl bromide, 98%, Thermo Scientific Chemicals
CAS: 589-15-1 | C7H6Br2 | 249.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number ACR150410250
Price (MYR)
485.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
485.00
Add to cart
Specifications
Chemical Name or Material4-Bromobenzyl bromide
Name Note98%
CAS589-15-1
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger