2,3-Butanedione monoxime, 98%, Thermo Scientific Chemicals
2,3-Butanedione monoxime, 98%, Thermo Scientific Chemicals
2,3-Butanedione monoxime, 98%, Thermo Scientific Chemicals
2,3-Butanedione monoxime, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,3-Butanedione monoxime, 98%, Thermo Scientific Chemicals
CAS: 57-71-6 | C4H7NO2 | 101.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
Plastic bottle
Catalog number ACR150375000
Price (MYR)
1,907.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,907.00
Add to cart
Chemical Identifiers
CAS7646-85-7
IUPAC Namezinc(2+) dichloride
Molecular FormulaCl2Zn
InChI KeyJIAARYAFYJHUJI-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Zn++]
View more
SpecificationsSpecification SheetSpecification Sheet
Water Content (Karl Fischer Titration)≤1.75%
FormCrystals or powder or crystalline powder
Appearance (Color)White to cream to pale grey
Assay (Titration ex Chloride)≥97.5 to ≤102.5%