2,3-Butanedione monoxime, 98%, Thermo Scientific Chemicals
2,3-Butanedione monoxime, 98%, Thermo Scientific Chemicals
2,3-Butanedione monoxime, 98%, Thermo Scientific Chemicals
2,3-Butanedione monoxime, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,3-Butanedione monoxime, 98%, Thermo Scientific Chemicals
CAS: 57-71-6 | C4H7NO2 | 101.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
Packaging:
Plastic bottle
Catalog number ACR150370025
Price (MYR)
7,623.00
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
7,623.00
Add to cart
Chemical Identifiers
CAS741709-63-7
IUPAC Name5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-2-carbonitrile
Molecular FormulaC12H15BN2O2
InChI KeyIXTBQKLZPOYJFJ-UHFFFAOYSA-N
SMILESCC1(C)OB(OC1(C)C)C1=CN=C(C=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White
Proton NMRConforms to structure