Hippuric acid, 98%, Thermo Scientific Chemicals
Hippuric acid, 98%, Thermo Scientific Chemicals
Hippuric acid, 98%, Thermo Scientific Chemicals
Hippuric acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hippuric acid, 98%, Thermo Scientific Chemicals
CAS: 495-69-2 | C9H9NO3 | 179.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
Packaging:
View all
Plastic bottle
Catalog number ACR150275000
Price (MYR)
698.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
698.00
Add to cart
Chemical Identifiers
CAS2942-06-5
IUPAC Name6-nitro-1,3-benzothiazole
Molecular FormulaC7H4N2O2S
InChI KeyQLUFBCVWKTWKBF-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C2N=CSC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (GC)≥98.0%
Melting Point (clear melt)171.5-177.5?C
Appearance (Color)Yellow to pale brown to pale cream