Hippuric acid, 98%, Thermo Scientific Chemicals
Hippuric acid, 98%, Thermo Scientific Chemicals
Hippuric acid, 98%, Thermo Scientific Chemicals
Hippuric acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hippuric acid, 98%, Thermo Scientific Chemicals
CAS: 495-69-2 | C9H9NO3 | 179.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
500 g
Packaging:
View all
Plastic bottle
Catalog number ACR150271000
Price (MYR)
407.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
407.00
Add to cart
Chemical Identifiers
CAS4463-44-9
IUPAC Namesodium 4-{[4-(ethylamino)-3-methylphenyl][(1Z)-4-(ethylimino)-3-methylcyclohexa-2,5-dien-1-ylidene]methyl}-6-hydroxybenzene-1,3-disulfonic acid hydride
Molecular FormulaC25H29N2NaO7S2
InChI KeyHLEQELPTFNOLFE-ZCTWALFCSA-N
SMILES[H-].[Na+].CCNC1=CC=C(C=C1C)C(=C1/C=CC(=NCC)C(C)=C1)\C1=CC(O)=C(C=C1S(O)(=O)=O)S(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Dye content>=80 %
Appearance (Color)Dark red to dark blue