Hippuric acid, 98%, Thermo Scientific Chemicals
Hippuric acid, 98%, Thermo Scientific Chemicals
Hippuric acid, 98%, Thermo Scientific Chemicals
Hippuric acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hippuric acid, 98%, Thermo Scientific Chemicals
CAS: 495-69-2 | C9H9NO3 | 179.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
View all
Glass Bottle
Catalog number ACR150270050
Price (MYR)
373.00
Add to cart
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
373.00
Add to cart
Specifications
Chemical Name or MaterialHippuric acid
Name Note98%
CAS495-69-2
Health Hazard 1GHS Signal Word: Danger
Melting Point188.0°C to 191.0°C