4-Chlorobenzyl chloride, 99+%, Thermo Scientific Chemicals
4-Chlorobenzyl chloride, 99+%, Thermo Scientific Chemicals
4-Chlorobenzyl chloride, 99+%, Thermo Scientific Chemicals
4-Chlorobenzyl chloride, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Chlorobenzyl chloride, 99+%, Thermo Scientific Chemicals
CAS: 104-83-6 | C7H6Cl2 | 161.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Catalog number ACR150242500
Price (MYR)
543.00
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
543.00
Add to cart
Chemical Identifiers
CAS1707-03-5
IUPAC Namediphenylphosphinic acid
Molecular FormulaC12H11O2P
InChI KeyBEQVQKJCLJBTKZ-UHFFFAOYSA-N
SMILESOP(=O)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Appearance (Color)White
Melting Point (clear melt)191.0-198.5°C
FormCrystals or powder or crystalline powder
Assay (GC)≥98.5% (non-U.S. sourced material)