n-Propyl Ether, 99+%
n-Propyl Ether, 99+%
n-Propyl Ether, 99+%
n-Propyl Ether, 99+%
Thermo Scientific Chemicals
n-Propyl Ether, 99+%
CAS: 111-43-3 | C6H14O | 102.18 g/mol
Have Questions?
Catalog number ACR150040500
Price (MYR)
1,179.00
Add to cart
Request bulk or custom format
Price (MYR)
1,179.00
Add to cart
Specifications
Chemical Name or Materialn-Propyl ether
Name Note99+%
CAS111-43-3
CAS Min %99.0
Health Hazard 1GHS Signal Word: Danger