2-Butanone, +99%, Extra Pure, Thermo Scientific Chemicals
2-Butanone, +99%, Extra Pure, Thermo Scientific Chemicals
2-Butanone, +99%, Extra Pure, Thermo Scientific Chemicals
2-Butanone, +99%, Extra Pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Butanone, +99%, Extra Pure, Thermo Scientific Chemicals
CAS: 78-93-3 | C4H8O | 72.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number ACR149670025
Price (MYR)
712.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
712.00
Add to cart
Chemical Identifiers
CAS536-38-9
IUPAC Name2-bromo-1-(4-chlorophenyl)ethan-1-one
Molecular FormulaC8H6BrClO
InChI KeyFLAYZKKEOIAALB-UHFFFAOYSA-N
SMILESClC1=CC=C(C=C1)C(=O)CBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
GC>=98.0 %
Appearance (Color)White to light yellow
Melting point95°C to 99°C