2-Butanone, +99%, Extra Pure, Thermo Scientific Chemicals
2-Butanone, +99%, Extra Pure, Thermo Scientific Chemicals
2-Butanone, +99%, Extra Pure, Thermo Scientific Chemicals
2-Butanone, +99%, Extra Pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Butanone, +99%, Extra Pure, Thermo Scientific Chemicals
CAS: 78-93-3 | C4H8O | 72.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number ACR149670010
Price (MYR)
619.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
619.00
Add to cart
Chemical Identifiers
CAS108-64-5
IUPAC Nameethyl 3-methylbutanoate
Molecular FormulaC7H14O2
InChI KeyPPXUHEORWJQRHJ-UHFFFAOYSA-N
SMILESCCOC(=O)CC(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Specific gravity(25/25°C) 0.861 to 0.865
Infrared spectrumConforms
Refractive index1.3950 to 1.3980 (20°C, 589 nm)
Appearance (Form)Liquid