Valeric Acid, 99%
Valeric Acid, 99%
Valeric Acid, 99%
Thermo Scientific Chemicals
Valeric Acid, 99%
CAS: 109-52-4 | C5H10O2 | 102.133 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
5 mL
2.5 L
Packaging:
Glass bottle
Catalog number ACR149571000
Price (MYR)
546.00
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
546.00
Add to cart
Chemical Identifiers
CAS108-80-5
IUPAC Name1,3,5-triazinane-2,4,6-trione
Molecular FormulaC3H3N3O3
InChI KeyZFSLODLOARCGLH-UHFFFAOYSA-N
SMILESO=C1NC(=O)NC(=O)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Appearance (Form)Crystalline powder and/or lumps
Infrared spectrumConforms
Titration with NaOH>=97.5 %