Valeric Acid, 99%
Valeric Acid, 99%
Valeric Acid, 99%
Thermo Scientific Chemicals
Valeric Acid, 99%
CAS: 109-52-4 | C5H10O2 | 102.133 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
100 mL
2.5 L
Packaging:
Glass bottle
Catalog number ACR149570050
Price (MYR)
558.00
Add to cart
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
558.00
Add to cart
Chemical Identifiers
CAS51388-20-6
IUPAC Namehydrogen 4-(benzyloxy)aniline chloride
Molecular FormulaC13H14ClNO
InChI KeyKQBDLOVXZHOAJI-UHFFFAOYSA-N
SMILES[H+].[Cl-].NC1=CC=C(OCC2=CC=CC=C2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to brown or pale grey to pale pink to pink
FormCrystals or powder or crystalline powder
Assay (Argentometric Titration)≥96.0 to ≤104.0%