Valeric Acid, 99%
Valeric Acid, 99%
Valeric Acid, 99%
Thermo Scientific Chemicals
Valeric Acid, 99%
CAS: 109-52-4 | C5H10O2 | 102.133 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
5 mL
100 mL
Packaging:
Glass bottle
Catalog number ACR149570025
Price (MYR)
1,103.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,103.00
Add to cart
Chemical Identifiers
CAS552-62-5
IUPAC Name7-methyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
Molecular FormulaC6H6N4O2
InChI KeyPFWLFWPASULGAN-UHFFFAOYSA-N
SMILESCN1C=NC2=C1C(=O)NC(=O)N2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to pale yellow or pale green
FormPowder
Assay (HPLC)≥97.5%