Acetonitrile, 99+%, extra pure, Thermo Scientific Chemicals
Acetonitrile, 99+%, extra pure, Thermo Scientific Chemicals
Acetonitrile, 99+%, extra pure, Thermo Scientific Chemicals
Acetonitrile, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acetonitrile, 99+%, extra pure, Thermo Scientific Chemicals
CAS: 75-05-8 | C2H3N | 41.053 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number ACR149520025
Price (MYR)
1,467.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,467.00
Add to cart
Chemical Identifiers
CAS19241-38-4
IUPAC Name4-bromo-1-isothiocyanato-2-methylbenzene
Molecular FormulaC8H6BrNS
InChI KeyYASXCQRGYJGIKD-UHFFFAOYSA-N
SMILESCC1=CC(Br)=CC=C1N=C=S
View more
SpecificationsSpecification SheetSpecification Sheet
FormFused solid
Assay (GC)≥97.5%
Appearance (Color)White to pale cream
Melting Point (clear melt)42.5-48.5?C