Acetonitrile, 99+%, extra pure, Thermo Scientific Chemicals
Acetonitrile, 99+%, extra pure, Thermo Scientific Chemicals
Acetonitrile, 99+%, extra pure, Thermo Scientific Chemicals
Acetonitrile, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acetonitrile, 99+%, extra pure, Thermo Scientific Chemicals
CAS: 75-05-8 | C2H3N | 41.053 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number ACR149520010
Price (MYR)
700.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
700.00
Add to cart
Chemical Identifiers
CAS193065-68-8
IUPAC Namezinc(2+) 3-ethoxy-3-oxopropan-1-ide bromide
Molecular FormulaC5H9BrO2Zn
InChI KeyAPPDFGHVJHZDKS-UHFFFAOYSA-M
SMILES[Zn++].[Br-].CCOC(=O)C[CH2-]
View more
SpecificationsSpecification SheetSpecification Sheet
Complexometric Titration0.40M - 0.60M
CommentMay form a precipitate on storage
Refractive Index1.4220-1.4300 @20?C
Appearance (Color)Brown to dark brown