1-Propanol, 99+%, Extra Pure, Thermo Scientific Chemicals
1-Propanol, 99+%, Extra Pure, Thermo Scientific Chemicals
1-Propanol, 99+%, Extra Pure, Thermo Scientific Chemicals
1-Propanol, 99+%, Extra Pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Propanol, 99+%, Extra Pure, Thermo Scientific Chemicals
CAS: 71-23-8 | C3H8O | 60.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number ACR149480025
Price (MYR)
895.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
895.00
Add to cart
Chemical Identifiers
CAS579-75-9
IUPAC Name2-methoxybenzoic acid
Molecular FormulaC8H8O3
InChI KeyILUJQPXNXACGAN-UHFFFAOYSA-N
SMILESCOC1=CC=CC=C1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
HPLC>=98.5 %
Appearance (Form)Crystalline powder
Melting point99°C to 103°C