1-Propanol, 99+%, Extra Pure, Thermo Scientific Chemicals
1-Propanol, 99+%, Extra Pure, Thermo Scientific Chemicals
1-Propanol, 99+%, Extra Pure, Thermo Scientific Chemicals
1-Propanol, 99+%, Extra Pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Propanol, 99+%, Extra Pure, Thermo Scientific Chemicals
CAS: 71-23-8 | C3H8O | 60.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number ACR149480010
Price (MYR)
556.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
556.00
Add to cart
Specifications
Chemical Name or Material1-Propanol
CAS71-23-8
CAS Min %99.0
Health Hazard 1Danger
CAS Max %100.0