Phenol, 99%, extra pure, Thermo Scientific Chemicals
Phenol, 99%, extra pure, Thermo Scientific Chemicals
Phenol, 99%, extra pure, Thermo Scientific Chemicals
Phenol, 99%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenol, 99%, extra pure, Thermo Scientific Chemicals
CAS: 108-95-2 | C6H6O | 94.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
1 kg
5 kg
Packaging:
View all
Glass bottle
Catalog number ACR149340500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialPhenol
Name Note99%
CAS108-95-2
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Toxic if inhaled.
Suspected of causing genetic defects.
May cause damage to organs through prolonged or repeated