Phenol, 99%, extra pure, Thermo Scientific Chemicals
Phenol, 99%, extra pure, Thermo Scientific Chemicals
Phenol, 99%, extra pure, Thermo Scientific Chemicals
Phenol, 99%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenol, 99%, extra pure, Thermo Scientific Chemicals
CAS: 108-95-2 | C6H6O | 94.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
50 g
5 kg
Packaging:
View all
Glass bottle
Catalog number ACR149340010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7440-22-4
IUPAC Namesilver
Molecular FormulaAg
InChI KeyBQCADISMDOOEFD-UHFFFAOYSA-N
SMILES[Ag]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥99.99% (metals basis)
Appearance (Color)Silver
FormFoil, 0.38mm (0.015in) thick.