Xanthine, 98%, Thermo Scientific Chemicals
Xanthine, 98%, Thermo Scientific Chemicals
Xanthine, 98%, Thermo Scientific Chemicals
Xanthine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Xanthine, 98%, Thermo Scientific Chemicals
CAS: 69-89-6 | C5H4N4O2 | 152.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number ACR149170250
Price (MYR)
1,090.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,090.00
Add to cart
Chemical Identifiers
CAS80-43-3
IUPAC Name{2-[(2-phenylpropan-2-yl)peroxy]propan-2-yl}benzene
Molecular FormulaC18H22O2
InChI KeyXMNIXWIUMCBBBL-UHFFFAOYSA-N
SMILESCC(C)(OOC(C)(C)C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals
Infrared spectrumConforms
Titration Iodometric>=98.5 %
Melting point38°C to 41°C
Appearance (Color)White