Iodoform, 99+%, Thermo Scientific Chemicals
Iodoform, 99+%, Thermo Scientific Chemicals
Iodoform, 99+%, Thermo Scientific Chemicals
Iodoform, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Iodoform, 99+%, Thermo Scientific Chemicals
CAS: 75-47-8 | CHI3 | 393.72 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number ACR149125000
Price (MYR)
1,945.00
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,945.00
Add to cart
Specifications
Chemical Name or MaterialIodoform
Name Note99+%
CAS75-47-8
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: