Iodoform, 99+%, Thermo Scientific Chemicals
Iodoform, 99+%, Thermo Scientific Chemicals
Iodoform, 99+%, Thermo Scientific Chemicals
Iodoform, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Iodoform, 99+%, Thermo Scientific Chemicals
CAS: 75-47-8 | CHI3 | 393.72 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number ACR149121000
Price (MYR)
671.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
671.00
Add to cart
Chemical Identifiers
CAS57235-50-4
SpecificationsSpecification SheetSpecification Sheet
Purity≥97.5%
Appearance (Color)Off-white to pale cream
Mass SpecConforms
FormPowder or crystals or crystalline powder