3-Heptanol, 98%, Thermo Scientific Chemicals
3-Heptanol, 98%, Thermo Scientific Chemicals
3-Heptanol, 98%, Thermo Scientific Chemicals
3-Heptanol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Heptanol, 98%, Thermo Scientific Chemicals
CAS: 589-82-2 | C7H16O | 116.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Catalog number ACR149090250
Price (MYR)
555.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
555.00
Add to cart
Specifications
Chemical Name or Material3-Heptanol
CAS589-82-2
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Flammable liquid and vapour.
Harmful if swallowed.
Health Hazard 3GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Store i