Aminoguanidine bicarbonate, 98.5%, Thermo Scientific Chemicals
Aminoguanidine bicarbonate, 98.5%, Thermo Scientific Chemicals
Aminoguanidine bicarbonate, 98.5%, Thermo Scientific Chemicals
Aminoguanidine bicarbonate, 98.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Aminoguanidine bicarbonate, 98.5%, Thermo Scientific Chemicals
CAS: 2582-30-1 | CH6N4·H2CO3 | 136.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Packaging:
Plastic Bottle
Catalog number ACR149062500
Price (MYR)
407.00
Add to cart
Quantity:
250 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
407.00
Add to cart
Specifications
Chemical Name or MaterialAminoguanidine bicarbonate
CAS2582-30-1
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic to aquatic life with long lasting effects.
May cause an allergic skin reaction.
May damage fertility or the unborn child.
Health Hazard 3GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention