Benzoic Acid, Sodium Salt, 99+%, Thermo Scientific Chemicals
Benzoic Acid, Sodium Salt, 99+%, Thermo Scientific Chemicals
Benzoic Acid, Sodium Salt, 99+%, Thermo Scientific Chemicals
Benzoic Acid, Sodium Salt, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Benzoic Acid, Sodium Salt, 99+%, Thermo Scientific Chemicals
CAS: 532-32-1 | C7H5NaO2 | 144.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
1 kg
Packaging:
View all
Plastic Drum
Catalog number ACR148980050
Price (MYR)
986.00
Add to cart
Quantity:
5 kg
Packaging:
Plastic Drum
Request bulk or custom format
Price (MYR)
986.00
Add to cart
Specifications
Chemical Name or MaterialSodium benzoate
CAS532-32-1
CAS Min %99.0
CAS Max %100.0