Benzoic Acid, Sodium Salt, 99+%, Thermo Scientific Chemicals
Benzoic Acid, Sodium Salt, 99+%, Thermo Scientific Chemicals
Benzoic Acid, Sodium Salt, 99+%, Thermo Scientific Chemicals
Benzoic Acid, Sodium Salt, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Benzoic Acid, Sodium Salt, 99+%, Thermo Scientific Chemicals
CAS: 532-32-1 | C7H5NaO2 | 144.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Packaging:
View all
Plastic Bottle
Catalog number ACR148980010
Price (MYR)
451.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
451.00
Add to cart
Chemical Identifiers
CAS1634-04-4
IUPAC Name2-methoxy-2-methylpropane
Molecular FormulaC5H12O
InChI KeyBZLVMXJERCGZMT-UHFFFAOYSA-N
SMILESCOC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
Water=<0.01 % (Coulometric)
Appearance (Form)Liquid
GC>=98.5 %