3,3'-Dithiodipropionic acid, 98%, Thermo Scientific Chemicals
3,3'-Dithiodipropionic acid, 98%, Thermo Scientific Chemicals
3,3'-Dithiodipropionic acid, 98%, Thermo Scientific Chemicals
3,3'-Dithiodipropionic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3,3'-Dithiodipropionic acid, 98%, Thermo Scientific Chemicals
CAS: 1119-62-6 | C6H10O4S2 | 210.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Packaging:
Glass bottle
Catalog number ACR148902500
Price (MYR)
897.00
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
897.00
Add to cart
Chemical Identifiers
CAS3178-22-1
IUPAC Nametert-butylcyclohexane
Molecular FormulaC10H20
InChI KeyXTVMZZBLCLWBPM-UHFFFAOYSA-N
SMILESCC(C)(C)C1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4460-1.4480 @ 20?C
FormLiquid
Assay (GC)≥99.0%
Appearance (Color)Clear colorless