3,3'-Dithiodipropionic acid, 98%, Thermo Scientific Chemicals
3,3'-Dithiodipropionic acid, 98%, Thermo Scientific Chemicals
3,3'-Dithiodipropionic acid, 98%, Thermo Scientific Chemicals
3,3'-Dithiodipropionic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3,3'-Dithiodipropionic acid, 98%, Thermo Scientific Chemicals
CAS: 1119-62-6 | C6H10O4S2 | 210.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Packaging:
Glass bottle
Catalog number ACR148900500
Price (MYR)
425.00
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
425.00
Add to cart
Chemical Identifiers
CAS1314-62-1
IUPAC Name[(dioxovanadio)oxy]dioxovanadium
Molecular FormulaO5V2
InChI KeyGNTDGMZSJNCJKK-UHFFFAOYSA-N
SMILESO=[V](=O)O[V](=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Sodium oxide (Na2O)=<300 ppm
Iron (Fe)=<1000 ppm
Chromium (Cr)=<200 ppm
Assay>=98.0 %