Mercaptoacetic acid, sodium salt, 98%, Thermo Scientific Chemicals
Mercaptoacetic acid, sodium salt, 98%, Thermo Scientific Chemicals
Mercaptoacetic acid, sodium salt, 98%, Thermo Scientific Chemicals
Mercaptoacetic acid, sodium salt, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Mercaptoacetic acid, sodium salt, 98%, Thermo Scientific Chemicals
CAS: 367-51-1 | C2H3NaO2S | 114.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Packaging:
Catalog number ACR148892500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS2028-63-9
IUPAC Namebut-3-yn-2-ol
Molecular FormulaC4H6O
InChI KeyGKPOMITUDGXOSB-UHFFFAOYNA-N
SMILESCC(O)C#C
View more
SpecificationsSpecification SheetSpecification Sheet
Optical Rotation0.0 ± 0.1? (neat)
Appearance (Color)Clear colorless to yellow
Refractive Index1.4235-1.4275 @ 20?C
Assay (GC)≥97.5%